Recombinat Protein,Biotinylated Human GITR TNFRSF18 Protein, Fc,Avitag,Biotinylated Human GITR, Fc,Avitag (GIR-H82F7) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Glu 161 (Accession Q9Y5U5-1).This protein carries a human IgG e 1 * 200 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Biotinylated Human GITR TNFRSF18 Protein, Fc,Avitag,Biotinylated Human GITR, Fc,Avitag (GIR-H82F7) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Glu 161 (Accession Q9Y5U5-1).This protein carries a human IgG e 1 * 200 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Biotinylated Human GITR TNFRSF18 Protein, Fc,Avitag™,Biotinylated Human GITR, Fc,Avitag (GIR-H82F7) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Glu 161 (Accession Q9Y5U5-1).This protein carries a human IgG e 1 * 200 µG ACRBGIR-H82F7200UG 1 * 200 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou