Biotinylated HIV-1 HIV-1Clade BC (CN54) GP120, His,Avitag derived from the env. gene of HIV-1 strain CN54 gp160 (Accession G4XFJ5-1 (E46G, T396A, A497T), Thr 36 - Arg 507) and glycosylated with N-linked sugars and expressed in HEK293 cells at ACRObi 1 * 200 µG
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Biotinylated HIV-1 HIV-1Clade BC (CN54) GP120, His,Avitag derived from the env. gene of HIV-1 strain CN54 gp160 (Accession G4XFJ5-1 (E46G, T396A, A497T), Thr 36 - Arg 507) and glycosylated with N-linked sugars and expressed in HEK293 cells at ACRObi 1 * 200 µG ACRBGP0V182E6200UG 1 * 200 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou