Human Glypican 1, His Tag (GP1-H52H9) is expressed from human 293 cells (HEK293). It contains AA Asp 24 - Ser 530 (Accession P35052 -1). 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Human Glypican 1, His Tag (GP1-H52H9) is expressed from human 293 cells (HEK293). It contains AA Asp 24 - Ser 530 (Accession P35052 -1). 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Human Glypican 1, His Tag (GP1-H52H9) is expressed from human 293 cells (HEK293). It contains AA Asp 24 - Ser 530 (Accession P35052 -1). 1 * 1 mg ACRBGP1-H52H9-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou