Recombinat Protein,Rhesus macaque Glypican 2 GPC2 Protein, His Tag,Rhesus macaque Glypican 2, His Tag (GP2-R52H9) is expressed from human 293 cells (HEK293). It contains AA Ser 24 - Ser 552 (Accession XP001103286).This protein carries a polyhistidi C 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Rhesus macaque Glypican 2 GPC2 Protein, His Tag,Rhesus macaque Glypican 2, His Tag (GP2-R52H9) is expressed from human 293 cells (HEK293). It contains AA Ser 24 - Ser 552 (Accession XP001103286).This protein carries a polyhistidi C 1 * 1 mg ACRBGP2-R52H9-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou