Cynomolgus Glypican 3, His Tag (GP3-C5225) is expressed from human 293 cells (HEK293). It contains AA Gln 25 - His 559 (Accession X P005594665.1). 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Cynomolgus Glypican 3, His Tag (GP3-C5225) is expressed from human 293 cells (HEK293). It contains AA Gln 25 - His 559 (Accession X P005594665.1). 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Cynomolgus Glypican 3, His Tag (GP3-C5225) is expressed from human 293 cells (HEK293). It contains AA Gln 25 - His 559 (Accession X P005594665.1). 1 * 1 mg ACRBGP3-C5225-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou