Recombinat Protein,Human GPA33 A33 Protein, His Tag,Human GPA33, His Tag (GP3-H5224) is expressed from human 293 cells (HEK293). It contains AA Ile 22 - Val 235 (Accession AAH74830).This protein carries a polyhistidine tag at the C-terminus. The pr l 1 * 50 µG
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human GPA33 A33 Protein, His Tag,Human GPA33, His Tag (GP3-H5224) is expressed from human 293 cells (HEK293). It contains AA Ile 22 - Val 235 (Accession AAH74830).This protein carries a polyhistidine tag at the C-terminus. The pr l 1 * 50 µG ACRBGP3-H5224-50UG 1 * 50 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou