MABSol® Biotinylated Human GPA33, His Tag, primary amine labeling (GP3-H8224) is expressed from human HEK293 cells. It contains AA I le 22 - Val 235 (Accession AAH74830). It is the biotinylated form of Human GPA33, His Tag (GP3-H5224). 1 * 200 µG
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
MABSol® Biotinylated Human GPA33, His Tag, primary amine labeling (GP3-H8224) is expressed from human HEK293 cells. It contains AA I le 22 - Val 235 (Accession AAH74830). It is the biotinylated form of Human GPA33, His Tag (GP3-H5224). 1 * 200 µG ACRBGP3-H8224200UG 1 * 200 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou