Biotinylated Human Glypican 3, His,Avitag (GP3-H82E5) is expressed from human 293 cells (HEK293). It contains AA Gln 25 - His 559 (A ccession P51654-1). 1 * 200 G
Fournisseur: ACROBIOSYSTEMS
Biotinylated Human Glypican 3, His,Avitag (GP3-H82E5) is expressed from human 293 cells (HEK293). It contains AA Gln 25 - His 559 (A ccession P51654-1). 1 * 200 G
COMMANDE
Développer tout / Réduire tout
Biotinylated Human Glypican 3, His,Avitag (GP3-H82E5) is expressed from human 293 cells (HEK293). It contains AA Gln 25 - His 559 (A ccession P51654-1). 1 * 200 µG ACRBGP3-H82E5200UG 1 * 200 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou