Recombinat Protein,Mouse GPA33 A33 Protein, His Tag,Mouse GPA33, His Tag (GP3-M5225) is expressed from human 293 cells (HEK293). It contains AA Leu 22 - Ile 235 (Accession NP067623.1).This protein carries a polyhistidine tag at the C-terminus. The ca 1 * 50 µG
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Mouse GPA33 A33 Protein, His Tag,Mouse GPA33, His Tag (GP3-M5225) is expressed from human 293 cells (HEK293). It contains AA Leu 22 - Ile 235 (Accession NP067623.1).This protein carries a polyhistidine tag at the C-terminus. The ca 1 * 50 µG ACRBGP3-M5225-50UG 1 * 50 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou