HIV-1 HIV-1Clade E (CM244) GP120, His Tag (GP4-V15223) is expressed from human 293 cells (HEK293). It contains AA Thr 36 - Lys 511 ( Accession Q4QX31). 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
HIV-1 HIV-1Clade E (CM244) GP120, His Tag (GP4-V15223) is expressed from human 293 cells (HEK293). It contains AA Thr 36 - Lys 511 ( Accession Q4QX31). 1 * 100 G
COMMANDE
Développer tout / Réduire tout
HIV-1 HIV-1Clade E (CM244) GP120, His Tag (GP4-V15223) is expressed from human 293 cells (HEK293). It contains AA Thr 36 - Lys 511 ( Accession Q4QX31). 1 * 100 µG ACRBGP4V15223100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou