HIV-1 HIV-1Clade C (16055) GP120, His Tag (GP5-V15224) is expressed from human 293 cells (HEK293). It contains AA Thr 34 - Arg 506 ( Accession ABL67444.1). 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
HIV-1 HIV-1Clade C (16055) GP120, His Tag (GP5-V15224) is expressed from human 293 cells (HEK293). It contains AA Thr 34 - Arg 506 ( Accession ABL67444.1). 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
HIV-1 HIV-1Clade C (16055) GP120, His Tag (GP5-V15224) is expressed from human 293 cells (HEK293). It contains AA Thr 34 - Arg 506 ( Accession ABL67444.1). 1 * 1 mg ACRBGP5-V15224-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou