Human GM-CSF R alpha, His Tag (SPR verified) (GRA-H52H7) is expressed from human 293 cells (HEK293). It contains AA Glu 23 - Gly 320 (Accession P15509-1). 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Human GM-CSF R alpha, His Tag (SPR verified) (GRA-H52H7) is expressed from human 293 cells (HEK293). It contains AA Glu 23 - Gly 320 (Accession P15509-1). 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Human GM-CSF R alpha, His Tag (SPR verified) (GRA-H52H7) is expressed from human 293 cells (HEK293). It contains AA Glu 23 - Gly 320 (Accession P15509-1). 1 * 1 mg ACRBGRA-H52H7-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou