Recombinat Protein,Human Serpin D1 Heparin Cofactor II Protein, His Tag,Human Serpin D1, His Tag (HC2-H5224) is expressed from huma n 293 cells (HEK293). It contains AA Gly 20 - Ser 499 (Accession AAH35028).This protein carries a polyhistidine tag a 1 * 50 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human Serpin D1 Heparin Cofactor II Protein, His Tag,Human Serpin D1, His Tag (HC2-H5224) is expressed from huma n 293 cells (HEK293). It contains AA Gly 20 - Ser 499 (Accession AAH35028).This protein carries a polyhistidine tag a 1 * 50 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human Serpin D1 Heparin Cofactor II Protein, His Tag,Human Serpin D1, His Tag (HC2-H5224) is expressed from huma n 293 cells (HEK293). It contains AA Gly 20 - Ser 499 (Accession AAH35028).This protein carries a polyhistidine tag a 1 * 50 µG ACRBHC2-H5224-50UG 1 * 50 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou