Cynomolgus Her2, His Tag (HE2-C52Hb) is expressed from human 293 cells (HEK293). It contains AA Thr 23 - Thr 652 (Accession XP00558 4091.2). 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Cynomolgus Her2, His Tag (HE2-C52Hb) is expressed from human 293 cells (HEK293). It contains AA Thr 23 - Thr 652 (Accession XP00558 4091.2). 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Cynomolgus Her2, His Tag (HE2-C52Hb) is expressed from human 293 cells (HEK293). It contains AA Thr 23 - Thr 652 (Accession XP00558 4091.2). 1 * 100 µG ACRBHE2-C52HB100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou