FITC-Labeled Human Her2, His Tag (HE2-HF224) is expressed from human 293 cells (HEK293). It contains AA Thr 23 - Thr 652 (Accession P04626-1). It is the FITC labeled form of Human Her2, His Tag (HE2-H5225). 1 * 200 µG
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
FITC-Labeled Human Her2, His Tag (HE2-HF224) is expressed from human 293 cells (HEK293). It contains AA Thr 23 - Thr 652 (Accession P04626-1). It is the FITC labeled form of Human Her2, His Tag (HE2-H5225). 1 * 200 µG ACRBHE2-HF224200UG 1 * 200 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou