Recombinat Protein,Human HE4 WFDC2 Protein, His Tag,Human HE4, His Tag (HE4-H5223) is expressed from human 293 cells (HEK293). It c ontains AA Glu 31 - Phe 124 (Accession AAH46106).This protein carries a polyhistidine tag at the C-terminus. The prot 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human HE4 WFDC2 Protein, His Tag,Human HE4, His Tag (HE4-H5223) is expressed from human 293 cells (HEK293). It c ontains AA Glu 31 - Phe 124 (Accession AAH46106).This protein carries a polyhistidine tag at the C-terminus. The prot 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human HE4 WFDC2 Protein, His Tag,Human HE4, His Tag (HE4-H5223) is expressed from human 293 cells (HEK293). It c ontains AA Glu 31 - Phe 124 (Accession AAH46106).This protein carries a polyhistidine tag at the C-terminus. The prot 1 * 1 mg ACRBHE4-H5223-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou