Recombinat Protein,Mouse HMGB1 Protein, His Tag,Mouse HMGB1, His Tag (HM1-M52H6) is expressed from human 293 cells (HEK293). It cont ains AA Met 1 - Glu 215 (Accession P63158-1).This protein carries a polyhistidine tag at the C-terminus. The protein 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Mouse HMGB1 Protein, His Tag,Mouse HMGB1, His Tag (HM1-M52H6) is expressed from human 293 cells (HEK293). It cont ains AA Met 1 - Glu 215 (Accession P63158-1).This protein carries a polyhistidine tag at the C-terminus. The protein 1 * 1 mg ACRBHM1-M52H6-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou