Recombinat Protein,Human HSP90AA1 HSP86 Protein, GST Tag,Human HSP90AA1, GST Tag (HS1-H513x) is expressed from E.coli cells. It con tains AA Glu 535 - Asp 732 (Accession NP005339).This protein carries a GST tag at the N-terminus. The protein has a c 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human HSP90AA1 HSP86 Protein, GST Tag,Human HSP90AA1, GST Tag (HS1-H513x) is expressed from E.coli cells. It con tains AA Glu 535 - Asp 732 (Accession NP005339).This protein carries a GST tag at the N-terminus. The protein has a c 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human HSP90AA1 HSP86 Protein, GST Tag,Human HSP90AA1, GST Tag (HS1-H513x) is expressed from E.coli cells. It con tains AA Glu 535 - Asp 732 (Accession NP005339).This protein carries a GST tag at the N-terminus. The protein has a c 1 * 100 µG ACRBHS1-H513X100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou