Human SOST, His Tag (HST-H5245) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Tyr 213 (Accession AAI01087.1). 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Human SOST, His Tag (HST-H5245) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Tyr 213 (Accession AAI01087.1). 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Human SOST, His Tag (HST-H5245) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Tyr 213 (Accession AAI01087.1). 1 * 100 µG ACRBHST-H5245100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou