Biotinylated Human ICOS (C136S, C137S), His,Avitag (recommended for biopanning) (ICS-H82E5) is expressed from human 293 cells (HEK29 3). It contains AA Glu 21 - Phe 141 (Accession Q9Y6W8-1 (C136S, C137S)). 1 * 200 G
Fournisseur: ACROBIOSYSTEMS
Biotinylated Human ICOS (C136S, C137S), His,Avitag (recommended for biopanning) (ICS-H82E5) is expressed from human 293 cells (HEK29 3). It contains AA Glu 21 - Phe 141 (Accession Q9Y6W8-1 (C136S, C137S)). 1 * 200 G
COMMANDE
Développer tout / Réduire tout
Biotinylated Human ICOS (C136S, C137S), His,Avitag (recommended for biopanning) (ICS-H82E5) is expressed from human 293 cells (HEK29 3). It contains AA Glu 21 - Phe 141 (Accession Q9Y6W8-1 (C136S, C137S)). 1 * 200 µG ACRBICS-H82E5200UG 1 * 200 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou