Mouse ICOS (C137S, C138S), His Tag (ICS-M52H8) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Leu 144 (Accessio n Q9WVS0-1 (C137S,C138S)). 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Mouse ICOS (C137S, C138S), His Tag (ICS-M52H8) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Leu 144 (Accessio n Q9WVS0-1 (C137S,C138S)). 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Mouse ICOS (C137S, C138S), His Tag (ICS-M52H8) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Leu 144 (Accessio n Q9WVS0-1 (C137S,C138S)). 1 * 1 mg ACRBICS-M52H8-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou