Human IFN-gamma R1, His Tag (IF1-H5223) is expressed from human 293 cells (HEK293). It contains AA Glu 18 - Gly 245 (Accession AAH 05333). 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Human IFN-gamma R1, His Tag (IF1-H5223) is expressed from human 293 cells (HEK293). It contains AA Glu 18 - Gly 245 (Accession AAH 05333). 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Human IFN-gamma R1, His Tag (IF1-H5223) is expressed from human 293 cells (HEK293). It contains AA Glu 18 - Gly 245 (Accession AAH 05333). 1 * 1 mg ACRBIF1-H5223-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou