Recombinat Protein,Human IFN-alpha beta R1 Protein, Fc Tag,Human IFNAR1, Fc Tag (IF1-H5253) is expressed from human 293 cells (HEK2 93). It contains AA Lys 28 - Lys 436 (Accession NP000620).This protein carries a human IgG1 Fc tag at the C-terminus. 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human IFN-alpha beta R1 Protein, Fc Tag,Human IFNAR1, Fc Tag (IF1-H5253) is expressed from human 293 cells (HEK2 93). It contains AA Lys 28 - Lys 436 (Accession NP000620).This protein carries a human IgG1 Fc tag at the C-terminus. 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human IFN-alpha beta R1 Protein, Fc Tag,Human IFNAR1, Fc Tag (IF1-H5253) is expressed from human 293 cells (HEK2 93). It contains AA Lys 28 - Lys 436 (Accession NP000620).This protein carries a human IgG1 Fc tag at the C-terminus. 1 * 1 mg ACRBIF1-H5253-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou