Recombinat Protein,Human IFN-alphabeta R2 Protein, His Tag,Human IFNAR2, His Tag (IF2-H5224) is expressed from human 293 cells (HEK2 93). It contains AA Ile 27 - Lys 243 (Accession NP997467).This protein carries a polyhistidine tag at the C-terminus 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human IFN-alphabeta R2 Protein, His Tag,Human IFNAR2, His Tag (IF2-H5224) is expressed from human 293 cells (HEK2 93). It contains AA Ile 27 - Lys 243 (Accession NP997467).This protein carries a polyhistidine tag at the C-terminus 1 * 1 mg ACRBIF2-H5224-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou