Recombinat Protein,Human IgG1 Fc Protein, His Tag,Human IgG Fc, His Tag (IG1-H5225) is expressed from human 293 cells (HEK293). It c ontains AA Glu 99 - Lys 330 (Accession P01857).This protein carries a polyhistidine tag at the C-terminus. The prote 1 * 200 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human IgG1 Fc Protein, His Tag,Human IgG Fc, His Tag (IG1-H5225) is expressed from human 293 cells (HEK293). It c ontains AA Glu 99 - Lys 330 (Accession P01857).This protein carries a polyhistidine tag at the C-terminus. The prote 1 * 200 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human IgG1 Fc Protein, His Tag,Human IgG Fc, His Tag (IG1-H5225) is expressed from human 293 cells (HEK293). It c ontains AA Glu 99 - Lys 330 (Accession P01857).This protein carries a polyhistidine tag at the C-terminus. The prote 1 * 200 µG ACRBIG1-H5225200UG 1 * 200 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou