Recombinat Protein,Human IgG3 Fc Protein, Tag Free,Human IgG Fc, Tag Free (IG3-H5200) is expressed from human 293 cells (HEK293). It contains AA Glu 99 - Lys 377 (Accession P01860).Human IgG3-Fc IGHG3 Region has a calculated MW of 31.2 kDa. The pr nu 1 * 200 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human IgG3 Fc Protein, Tag Free,Human IgG Fc, Tag Free (IG3-H5200) is expressed from human 293 cells (HEK293). It contains AA Glu 99 - Lys 377 (Accession P01860).Human IgG3-Fc IGHG3 Region has a calculated MW of 31.2 kDa. The pr nu 1 * 200 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human IgG3 Fc Protein, Tag Free,Human IgG Fc, Tag Free (IG3-H5200) is expressed from human 293 cells (HEK293). It contains AA Glu 99 - Lys 377 (Accession P01860).Human IgG3-Fc IGHG3 Region has a calculated MW of 31.2 kDa. The pr nu 1 * 200 µG ACRBIG3-H5200200UG 1 * 200 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou