Recombinat Protein,Human IGFBP-7 Protein, His Tag,Human IGFBP-7, His Tag (IG7-H5240) is expressed from human 293 cells (HEK293). It contains AA Asp 30 - Leu 282 (Accession NP001544).This protein carries a polyhistidine tag at the N-terminus. The pro 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human IGFBP-7 Protein, His Tag,Human IGFBP-7, His Tag (IG7-H5240) is expressed from human 293 cells (HEK293). It contains AA Asp 30 - Leu 282 (Accession NP001544).This protein carries a polyhistidine tag at the N-terminus. The pro 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human IGFBP-7 Protein, His Tag,Human IGFBP-7, His Tag (IG7-H5240) is expressed from human 293 cells (HEK293). It contains AA Asp 30 - Leu 282 (Accession NP001544).This protein carries a polyhistidine tag at the N-terminus. The pro 1 * 1 mg ACRBIG7-H5240-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou