Recombinat Protein,Human IGFBP-7 Protein, His Tag,Human IGFBP-7, His Tag (IG7-H5240) is expressed from human 293 cells (HEK293). It contains AA Asp 30 - Leu 282 (Accession NP001544).This protein carries a polyhistidine tag at the N-terminus. The pro 1 * 50 µG
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human IGFBP-7 Protein, His Tag,Human IGFBP-7, His Tag (IG7-H5240) is expressed from human 293 cells (HEK293). It contains AA Asp 30 - Leu 282 (Accession NP001544).This protein carries a polyhistidine tag at the N-terminus. The pro 1 * 50 µG ACRBIG7-H5240-50UG 1 * 50 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou