Recombinat Protein,Mouse IgG2a Fc Protein, Tag Free,Mouse IgG Fc, Tag Free (IGA-M5207) is expressed from human 293 cells (HEK293). I t contains AA Glu 98 - Lys 330 (Accession CAC20702).This protein carries no tag. The protein has a calculated MW of r 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Mouse IgG2a Fc Protein, Tag Free,Mouse IgG Fc, Tag Free (IGA-M5207) is expressed from human 293 cells (HEK293). I t contains AA Glu 98 - Lys 330 (Accession CAC20702).This protein carries no tag. The protein has a calculated MW of r 1 * 1 mg ACRBIGA-M5207-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou