Cynomolgus IGF-I R, His Tag (IGR-C5225) is expressed from human 293 cells (HEK293). It contains AA Glu 31 - Asn 932 (Accession G7P9 I7-1). 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Cynomolgus IGF-I R, His Tag (IGR-C5225) is expressed from human 293 cells (HEK293). It contains AA Glu 31 - Asn 932 (Accession G7P9 I7-1). 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Cynomolgus IGF-I R, His Tag (IGR-C5225) is expressed from human 293 cells (HEK293). It contains AA Glu 31 - Asn 932 (Accession G7P9 I7-1). 1 * 100 µG ACRBIGR-C5225100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou