Human IGF-I R, His Tag (IGR-H5229) is expressed from human 293 cells (HEK293). It contains AA Glu 31 - Asn 932 (Accession P08069-1) . 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Human IGF-I R, His Tag (IGR-H5229) is expressed from human 293 cells (HEK293). It contains AA Glu 31 - Asn 932 (Accession P08069-1) . 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Human IGF-I R, His Tag (IGR-H5229) is expressed from human 293 cells (HEK293). It contains AA Glu 31 - Asn 932 (Accession P08069-1) . 1 * 100 µG ACRBIGR-H5229100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou