Recombinat Protein,Human IL-10 Protein, His Tag,Human IL-10, His Tag (IL0-H4248) is expressed from human 293 cells (HEK293). It cont ains AA Ser 19 - Asn 178 (Accession NP000563.1).This protein carries a polyhistidine tag at the N-terminus. The prot 1 * 20 µG
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human IL-10 Protein, His Tag,Human IL-10, His Tag (IL0-H4248) is expressed from human 293 cells (HEK293). It cont ains AA Ser 19 - Asn 178 (Accession NP000563.1).This protein carries a polyhistidine tag at the N-terminus. The prot 1 * 20 µG ACRBIL0-H4248-20UG 1 * 20 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou