Recombinat Protein,Human IL-1 RII CD121b Protein, His Tag,Human IL-1 RII, His Tag (IL2-H4226) is expressed from human 293 cells (HE K293). It contains AA Thr 14 - Glu 343 (Accession NP004624.1).This protein carries a polyhistidine tag at the C-termi 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human IL-1 RII CD121b Protein, His Tag,Human IL-1 RII, His Tag (IL2-H4226) is expressed from human 293 cells (HE K293). It contains AA Thr 14 - Glu 343 (Accession NP004624.1).This protein carries a polyhistidine tag at the C-termi 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human IL-1 RII CD121b Protein, His Tag,Human IL-1 RII, His Tag (IL2-H4226) is expressed from human 293 cells (HE K293). It contains AA Thr 14 - Glu 343 (Accession NP004624.1).This protein carries a polyhistidine tag at the C-termi 1 * 1 mg ACRBIL2-H4226-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou