Human IL-22, His Tag (IL2-H524a) is expressed from human 293 cells (HEK293). It contains AA Ala 34 - Ile 179 (Accession Q9GZX6-1). 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Human IL-22, His Tag (IL2-H524a) is expressed from human 293 cells (HEK293). It contains AA Ala 34 - Ile 179 (Accession Q9GZX6-1). 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Human IL-22, His Tag (IL2-H524a) is expressed from human 293 cells (HEK293). It contains AA Ala 34 - Ile 179 (Accession Q9GZX6-1). 1 * 1 mg ACRBIL2-H524A-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou