Human IL-2, His Tag (IL2-H52H8) is expressed from human 293 cells (HEK293). It contains AA Ala 21 - Thr 153 (Accession P60568-1). 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Human IL-2, His Tag (IL2-H52H8) is expressed from human 293 cells (HEK293). It contains AA Ala 21 - Thr 153 (Accession P60568-1). 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Human IL-2, His Tag (IL2-H52H8) is expressed from human 293 cells (HEK293). It contains AA Ala 21 - Thr 153 (Accession P60568-1). 1 * 100 µG ACRBIL2-H52H8100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou