Human IL-33, His Tag (IL3-H52H7) is expressed from human 293 cells (HEK293). It contains AA His 109 - Thr 270 (Accession O95760-1). 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Human IL-33, His Tag (IL3-H52H7) is expressed from human 293 cells (HEK293). It contains AA His 109 - Thr 270 (Accession O95760-1). 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Human IL-33, His Tag (IL3-H52H7) is expressed from human 293 cells (HEK293). It contains AA His 109 - Thr 270 (Accession O95760-1). 1 * 1 mg ACRBIL3-H52H7-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou