ActiveMax Human IL-4, Tag Free (IL4-H4218) is expressed from human 293 cells (HEK293). It contains AA His 25 - Ser 153 (Accession P05112-1). 1 * 20 G
Fournisseur: ACROBIOSYSTEMS
ActiveMax Human IL-4, Tag Free (IL4-H4218) is expressed from human 293 cells (HEK293). It contains AA His 25 - Ser 153 (Accession P05112-1). 1 * 20 G
COMMANDE
Développer tout / Réduire tout
ActiveMax® Human IL-4, Tag Free (IL4-H4218) is expressed from human 293 cells (HEK293). It contains AA His 25 - Ser 153 (Accession P05112-1). 1 * 20 µG ACRBIL4-H4218-20UG 1 * 20 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou