Biotinylated Human IL-4, Avitag,His Tag (IL4-H82E0) is expressed from human 293 cells (HEK293). It contains AA His 25 - Ser 153 (Acc ession AAH67514). 1 * 25 G
Fournisseur: ACROBIOSYSTEMS
Biotinylated Human IL-4, Avitag,His Tag (IL4-H82E0) is expressed from human 293 cells (HEK293). It contains AA His 25 - Ser 153 (Acc ession AAH67514). 1 * 25 G
COMMANDE
Développer tout / Réduire tout
Biotinylated Human IL-4, Avitag,His Tag (IL4-H82E0) is expressed from human 293 cells (HEK293). It contains AA His 25 - Ser 153 (Acc ession AAH67514). 1 * 25 µG ACRBIL4-H82E0-25UG 1 * 25 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou