Human IL-5, His Tag (IL5-H52H3) is expressed from human 293 cells (HEK293). It contains AA Ile 20 - Ser 134 (Accession P05113-1). 1 * 50 G
Fournisseur: ACROBIOSYSTEMS
Human IL-5, His Tag (IL5-H52H3) is expressed from human 293 cells (HEK293). It contains AA Ile 20 - Ser 134 (Accession P05113-1). 1 * 50 G
COMMANDE
Développer tout / Réduire tout
Human IL-5, His Tag (IL5-H52H3) is expressed from human 293 cells (HEK293). It contains AA Ile 20 - Ser 134 (Accession P05113-1). 1 * 50 µG ACRBIL5-H52H3-50UG 1 * 50 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou