MABSol Biotinylated Human IL-6, epitope tag free, primary amine labeling (IL6-H8218) is expressed from human HEK293 cells. It cont ains AA Val 30 - Met 212 (Accession NP000591). It is the biotinylated form of ActiveMax Human IL-6, Tag Free (IL6-H 1 * 200 G
Fournisseur: ACROBIOSYSTEMS
MABSol Biotinylated Human IL-6, epitope tag free, primary amine labeling (IL6-H8218) is expressed from human HEK293 cells. It cont ains AA Val 30 - Met 212 (Accession NP000591). It is the biotinylated form of ActiveMax Human IL-6, Tag Free (IL6-H 1 * 200 G
COMMANDE
Développer tout / Réduire tout
MABSol® Biotinylated Human IL-6, epitope tag free, primary amine labeling (IL6-H8218) is expressed from human HEK293 cells. It cont ains AA Val 30 - Met 212 (Accession NP000591). It is the biotinylated form of ActiveMax® Human IL-6, Tag Free (IL6-H 1 * 200 µG ACRBIL6-H8218200UG 1 * 200 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou