ActiveMax Human IL-7, Tag Free (IL7-H4219) is expressed from human 293 cells (HEK293). It contains AA Asp 26 - His 177 (Accession P13232). 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
ActiveMax Human IL-7, Tag Free (IL7-H4219) is expressed from human 293 cells (HEK293). It contains AA Asp 26 - His 177 (Accession P13232). 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
ActiveMax® Human IL-7, Tag Free (IL7-H4219) is expressed from human 293 cells (HEK293). It contains AA Asp 26 - His 177 (Accession P13232). 1 * 1 mg ACRBIL7-H4219-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou