Recombinat Protein,Human IL-8 CXCL8 Protein, His Tag,Human IL-8, His Tag (IL8-H52H3) is expressed from human 293 cells (HEK293). It contains AA Ser 28 - Ser 99 (Accession P10145-1).This protein carries a polyhistidine tag at the C-terminus. The pro c 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human IL-8 CXCL8 Protein, His Tag,Human IL-8, His Tag (IL8-H52H3) is expressed from human 293 cells (HEK293). It contains AA Ser 28 - Ser 99 (Accession P10145-1).This protein carries a polyhistidine tag at the C-terminus. The pro c 1 * 1 mg ACRBIL8-H52H3-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou