Recombinat Protein,Human IL-29 IFN-lambda 1 Protein, His Tag,Human IL-29, His Tag (IL9-H4228) is expressed from human 293 cells (HE K293). It contains AA Met 1 - Thr 200 (Accession NP742152.1).This protein carries a polyhistidine tag at the C-termin 1 * 50 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human IL-29 IFN-lambda 1 Protein, His Tag,Human IL-29, His Tag (IL9-H4228) is expressed from human 293 cells (HE K293). It contains AA Met 1 - Thr 200 (Accession NP742152.1).This protein carries a polyhistidine tag at the C-termin 1 * 50 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human IL-29 IFN-lambda 1 Protein, His Tag,Human IL-29, His Tag (IL9-H4228) is expressed from human 293 cells (HE K293). It contains AA Met 1 - Thr 200 (Accession NP742152.1).This protein carries a polyhistidine tag at the C-termin 1 * 50 µG ACRBIL9-H4228-50UG 1 * 50 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou