Recombinat Protein,Cynomolgus IL-3 R alpha CD123 Protein, His Tag,Cynomolgus IL-3 R alpha, His Tag (ILA-C52H6) is expressed from hu man 293 cells (HEK293). It contains AA Arg 18 - Arg 302 (Accession G8F3K3-1).This protein carries a polyhistidine tag 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Cynomolgus IL-3 R alpha CD123 Protein, His Tag,Cynomolgus IL-3 R alpha, His Tag (ILA-C52H6) is expressed from hu man 293 cells (HEK293). It contains AA Arg 18 - Arg 302 (Accession G8F3K3-1).This protein carries a polyhistidine tag 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Cynomolgus IL-3 R alpha CD123 Protein, His Tag,Cynomolgus IL-3 R alpha, His Tag (ILA-C52H6) is expressed from hu man 293 cells (HEK293). It contains AA Arg 18 - Arg 302 (Accession G8F3K3-1).This protein carries a polyhistidine tag 1 * 100 µG ACRBILA-C52H6100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou