Recombinat Protein,Human IL-5 R alpha CD125 Protein, His Tag,Human IL-5 R alpha, His Tag (ILA-H52H4) is expressed from human 293 ce lls (HEK293). It contains AA Asp 21 - Glu 335 (Accession NP000555).This protein carries a polyhistidine tag at the C- 1 * 50 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human IL-5 R alpha CD125 Protein, His Tag,Human IL-5 R alpha, His Tag (ILA-H52H4) is expressed from human 293 ce lls (HEK293). It contains AA Asp 21 - Glu 335 (Accession NP000555).This protein carries a polyhistidine tag at the C- 1 * 50 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human IL-5 R alpha CD125 Protein, His Tag,Human IL-5 R alpha, His Tag (ILA-H52H4) is expressed from human 293 ce lls (HEK293). It contains AA Asp 21 - Glu 335 (Accession NP000555).This protein carries a polyhistidine tag at the C- 1 * 50 µG ACRBILA-H52H4-50UG 1 * 50 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou