Human IL-2 R alpha, His Tag (ILA-H52H9) is expressed from human 293 cells (HEK293). It contains AA Glu 22 - Cys 213 (Accession NP00 0408). 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Human IL-2 R alpha, His Tag (ILA-H52H9) is expressed from human 293 cells (HEK293). It contains AA Glu 22 - Cys 213 (Accession NP00 0408). 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Human IL-2 R alpha, His Tag (ILA-H52H9) is expressed from human 293 cells (HEK293). It contains AA Glu 22 - Cys 213 (Accession NP00 0408). 1 * 100 µG ACRBILA-H52H9100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou