Recombinat Protein,Human IL-37 IL-1F7b Protein, His Tag,Human IL-37, His Tag (ILB-H5141) is expressed from E.coli cells. It contain s AA Val 46 - Asp 218 (Accession AAH20637).This protein carries a polyhistidine tag at the N-terminus. The protein ha 1 * 50 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human IL-37 IL-1F7b Protein, His Tag,Human IL-37, His Tag (ILB-H5141) is expressed from E.coli cells. It contain s AA Val 46 - Asp 218 (Accession AAH20637).This protein carries a polyhistidine tag at the N-terminus. The protein ha 1 * 50 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human IL-37 IL-1F7b Protein, His Tag,Human IL-37, His Tag (ILB-H5141) is expressed from E.coli cells. It contain s AA Val 46 - Asp 218 (Accession AAH20637).This protein carries a polyhistidine tag at the N-terminus. The protein ha 1 * 50 µG ACRBILB-H5141-50UG 1 * 50 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou