Recombinat Protein,Human IL-18BP Protein, His Tag,Human IL-18BP, His Tag (ILP-H5222) is expressed from human 293 cells (HEK293). It contains AA Thr 31 - Gly 194 (Accession NP001138529).This protein carries a polyhistidine tag at the C-terminus. The c 1 * 200 µG
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human IL-18BP Protein, His Tag,Human IL-18BP, His Tag (ILP-H5222) is expressed from human 293 cells (HEK293). It contains AA Thr 31 - Gly 194 (Accession NP001138529).This protein carries a polyhistidine tag at the C-terminus. The c 1 * 200 µG ACRBILP-H5222200UG 1 * 200 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou