Human IL-4 R alpha, His Tag (ILR-H5221) is expressed from human 293 cells (HEK293). It contains AA Met 26 - His 232 (Accession NP00 0409.1). 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Human IL-4 R alpha, His Tag (ILR-H5221) is expressed from human 293 cells (HEK293). It contains AA Met 26 - His 232 (Accession NP00 0409.1). 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Human IL-4 R alpha, His Tag (ILR-H5221) is expressed from human 293 cells (HEK293). It contains AA Met 26 - His 232 (Accession NP00 0409.1). 1 * 1 mg ACRBILR-H5221-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou