Recombinat Protein,Mouse IL-4 R alpha CD124 Protein, His Tag,Mouse IL-4 R alpha, His Tag (ILR-M52H1) is expressed from human 293 ce lls (HEK293). It contains AA Ile 26 - Arg 233 (Accession NP001008700).This protein carries a polyhistidine tag at the 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Mouse IL-4 R alpha CD124 Protein, His Tag,Mouse IL-4 R alpha, His Tag (ILR-M52H1) is expressed from human 293 ce lls (HEK293). It contains AA Ile 26 - Arg 233 (Accession NP001008700).This protein carries a polyhistidine tag at the 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Mouse IL-4 R alpha CD124 Protein, His Tag,Mouse IL-4 R alpha, His Tag (ILR-M52H1) is expressed from human 293 ce lls (HEK293). It contains AA Ile 26 - Arg 233 (Accession NP001008700).This protein carries a polyhistidine tag at the 1 * 1 mg ACRBILR-M52H1-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou